Skip to Content
Merck

1.46326

Tryptone Bile X-Glucuronide Agar

ReadyPlate, plate diam. 90 mm, ready-to-use, sterile, box of 20 plates, for Escherichia coli

Synonym(s):

ReadyPlate CHROM TBX, ReadyPlate Tryptone Bile X-Glucuronide Agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41106217
Plate diam.:
90 mm
Feature:
ready-to-use
Sterility:
sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

ISO 16649

Quality Segment

sterility

sterile

form

plate

feature

ready-to-use

packaging

box of 20 plates

manufacturer/tradename

ReadyPlate

plate diam.

90 mm

pH

7.2 ( in H2O)

application(s)

food and beverages
microbiology
water monitoring

storage temp.

2-8°C

suitability

Escherichia coli

General description

ReadyPlate Tryptone Bile X-Glucuronide Agar plates are ready-to-use, sterile 90mm plates for the enumeration, detection, and identification of β-glucuronidase positive Escherichia coli from food and feeding stuff. The presence of the enzyme β-D-glucuronidase differentiates most Escherichia coli from other coliforms. E. coli absorbs the chromogenic substrate 5-bromo-4-chloro-3-indolyl-β-D glucuronide (X-β-D-glucuronide). The enzyme β-glucuronidase splits the bond between the chromophore 5-bromo-4-chloro-3-indolyle- and the β-D-glucuronide forming blue to blue-green colored colonies. The growth of accompanying Gram-positive flora is largely inhibited by the use of bile salts and the high incubation temperature of 44°C.

Application

Tryptone Bile X-Glucuronide Agar plates are ready-to-use, sterile 90mm plates for the enumeration, detection, and identification of β-glucuronidase positive Escherichia coli from food and feeding stuff as well as from environmental samples in the area of food production and food handling.

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Tryptone Bile X-Glucuronide Agar


Storage Class

11 - Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library