Skip to Content
Merck

1.37038

Phenol red (phenolsulfonphthaleine)

EMPROVE® ESSENTIAL Ph Eur

Synonym(s):

Phenol red (phenolsulfonphthaleine)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C6H4OH)2C7H4SO3
CAS Number:
Molecular Weight:
354.38
UNSPSC Code:
12171502
EC Index Number:
205-609-7
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

Ph. Eur.

Quality Segment

assay

≥98.0% (HPLC)

form

solid

mp

>300 °C

solubility

0.77 g/L

bulk density

200‑300 kg/m3

storage temp.

2-30°C

SMILES string

[S]1(=O)(=O)OC(c4c1cccc4)(c3ccc(cc3)O)c2ccc(cc2)O

InChI

1S/C19H14O5S/c20-15-9-5-13(6-10-15)19(14-7-11-16(21)12-8-14)17-3-1-2-4-18(17)25(22,23)24-19/h1-12,20-21H

InChI key

BELBBZDIHDAJOR-UHFFFAOYSA-N

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library