Skip to Content
Merck

A-191

Apixaban solution

1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H25N5O4
CAS Number:
Molecular Weight:
459.50
UNSPSC Code:
12352107
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1=NN(C2=C1CCN(C3=CC=C(N4C(CCCC4)=O)C=C3)C2=O)C5=CC=C(C=C5)OC)N

InChI

1S/C25H25N5O4/c1-34-19-11-9-18(10-12-19)30-23-20(22(27-30)24(26)32)13-15-29(25(23)33)17-7-5-16(6-8-17)28-14-3-2-4-21(28)31/h5-12H,2-4,13-15H2,1H3,(H2,26,32)

InChI key

QNZCBYKSOIHPEH-UHFFFAOYSA-N

General description

This Certified Spiking Solution® is suitable as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from therapeutic drug monitoring, pharmaceutical research, forensic analysis and clinical toxicology to urine drug testing. Apixaban, sold as Eliquis, is an anticoagulant used to treat and prevent blood clots and to prevent stroke.

Legal Information

Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library