Skip to Content
Merck
All Photos(2)

Key Documents

858144P

Avanti

18:0 Lyso PS

1-stearoyl-2-hydroxy-sn-glycero-3-phospho-L-serine (sodium salt), powder

Synonym(s):

1-octadecanoyl-sn-glycero-3-phospho-L-serine (sodium salt); 18:0 LPS; C18 LPS; PS(18:0/0:0); 110725

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H47NPO9Na
CAS Number:
Molecular Weight:
547.59
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

99% (LPS; may contain up to 15% of the 2-LPS isomer, TLC)

form

powder

packaging

pkg of 1 × 1 mg (858144P-1mg)
pkg of 1 × 5 mg (858144P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 858144P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(O)COC(CCCCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C24H48NO9P.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-23(27)32-18-21(26)19-33-35(30,31)34-20-22(25)24(28)29;/h21-22,26H,2-20,25H2,1H3,(H,28,29)(H,30,31);/q;+1/p-1/t21-,22+;/m1./s1

InChI key

SVSJEWKLQKWQOV-NSLUPJTDSA-M

Application

18:0 Lyso PS has been used to stimulate the increase in intracellular cyclic adenosine monophosphate (cAMP) in G protein-coupled receptor 174 (GPR174)-cells. It may also be used in liquid chromatography-mass spectrometry (LC-MS) based structural characterization of oxygenated/hydrolyzed molecular species of phosphatidylserine (PS).

Packaging

5 mL Amber Glass Screw Cap Vial (858144P-1mg)
5 mL Amber Glass Screw Cap Vial (858144P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vladimir A Tyurin et al.
Biochemistry, 51(48), 9736-9750 (2012-11-15)
Ca(2+)-independent lipoprotein-associated phospholipase A(2) (Lp-PLA(2)) is a member of the phospholipase A(2) superfamily with a distinguishing characteristic of high specificity for oxidatively modified sn-2 fatty acid residues in phospholipids that has been especially well characterized for peroxidized species of phosphatidylcholines
Kazuya Sugita et al.
Biochemical and biophysical research communications, 430(1), 190-195 (2012-11-28)
We established cell lines that stably express orphan GPCR GPR174 using CHO cells, and studied physiological and pharmacological features of the receptor. GPR174-expressing cells showed cell-cell adhesion with localization of actin filaments to cell membrane, and revealed significant delay of
Ming Zhu et al.
International immunopharmacology, 81, 106034-106034 (2019-12-02)
GPR174 plays a crucial role in immune responses, but the role of GPR174 in the pathological progress of sepsis remains incompletely understood. In this study, we generated a sepsis model by cecal ligation and puncture (CLP) to investigate the role
Masako Nishikawa et al.
Journal of atherosclerosis and thrombosis, 22(5), 518-526 (2014-12-03)
Lysophospholipids, particularly sphingosine 1-phosphate and lysophosphatidic acid, are known to be involved in the pathogenesis of atherosclerosis; however, the role of lysophosphatidylserine (LysoPS) in the onset of atherosclerotic diseases remains uncertain. We investigated the effects of LysoPS on the uptake

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service