Skip to Content
Merck
All Photos(1)

Key Documents

SLW1521/08D

Pyrex® Reagent bottles, narrow neck with stopper

capacity 500 mL

Synonym(s):

Pyrex, SciLabware 1521/08D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

material

Pyrex bottle
colorless bottle
glass cap
glass stopper
narrow-mouth bottle

Joint

joint ST/NS neck24/29

feature

with cap

packaging

pack of 10 ea

manufacturer/tradename

1521/08D
SciLabware 1521/08D
SciLabware

bottle diam. × H

89 mm × 175 mm , With stopper

capacity

500 mL

neck I.D.

24 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • Complies with ISO 4796-2
  • Borosilicate glass with ground neck
  • Supplied with solid glass stopper

Features and Benefits

  • Borosilicate glass
  • Complies with ISO 4796-2
  • With ground neck
  • Supplied with solid glass stopper
  • Spare replacement glass stoppers are available

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service