20351
ORBO™ 60 FLORISIL® purified/deactivated (30/45), 100/50 mg
W,F,F (specially cleaned) separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
About This Item
Recommended Products
material
W,F,F (specially cleaned) separators
Agency
NIOSH 5503
product line
ORBO™
composition
Bed A, 100 mg
Bed B, 50 mg
packaging
pkg of 50 ea
manufacturer/tradename
ORBO™ 60
technique(s)
active air sampling: suitable
O.D. × L
6 mm × 75 mm
matrix
Florisil® magnesium silicate (purified/deactivated)
particle size
30-45 mesh
application(s)
air monitoring
environmental
industrial hygiene
General description
- ORBO™ 60 FLORISIL® purified/deactivated (30/45), 100/50 mg solvent desorption tube is designed for gas and vapor sampling.
- It contains two beds of the same selective adsorbent separated by glass wool or foam.
- Dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service