Merck
TMO4112-2000
Nalgene® sterile single-use Erlenmeyer flask with solid closure
PETG, bottle volume 2000 mL, case of 4 ea
Synonym(s):
Nalgene Erlenmeyer flask, single use plastic flask, Nalgene 4112-2000, disposable flask, plastic flask
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.13
UNSPSC Code:
41121804
Material:
PETG
Sterility:
sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
PETG
sterility
sterile
packaging
case of 4 ea
manufacturer/tradename
Nalgene 4112-2000
bottle volume
2000 mL
General description
Save preparation time and avoid contamination risk by using Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Other Notes
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)