Key Documents
T1068
Pharmed L/S tubing
size 18
Synonym(s):
Saint Gobain AY242022, Peristaltic Pump Tubing
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
packaging
pack of 25 feet
manufacturer/tradename
Tygon
I.D.
0.31 in. (7.9 mm)
General description
Tubing for MasterFlex Peristaltic Pumps, PharMed tubing has been specifically formulated for the MasterFlex pump system. Great for tissue and cell work- nontoxic and nonhemolytic. Long service life minimizes risk of fluid exposure; reduces tubing cost and pump downtime.
  • Opaque to UV and visible light to protect light-sensitive fluids
  • Heat sealable, bondable, and formable
  • Extremely low gas permeability
Legal Information
MasterFlex is a trademark of Cole-Parmer Instrument Co.