Skip to Content
Merck
All Photos(1)

Key Documents

SML1844

Sigma-Aldrich

EPI-001

≥98% (HPLC)

Synonym(s):

3-[4-[1-[4-(3-Chloro-2-hydroxypropoxy)phenyl]-1-methylethyl]phenoxy]-1,2-propanediol, EPI 001

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27ClO5
CAS Number:
Molecular Weight:
394.89
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

SMILES string

ClCC(O)COc1ccc(cc1)C(C)(C)c2ccc(cc2)OCC(O)CO

InChI

1S/C21H27ClO5/c1-21(2,15-3-7-19(8-4-15)26-13-17(24)11-22)16-5-9-20(10-6-16)27-14-18(25)12-23/h3-10,17-18,23-25H,11-14H2,1-2H3

InChI key

HDTYUHNZRYZEEB-UHFFFAOYSA-N

Biochem/physiol Actions

EPI-001 is a specific inhibitor of the AR (androgen receptor) without inhibiting PR or GR transcriptional activities. EPI-001 selectively interacts with transactivation unit 5 (Tau-5) of the androgen receptor, which is essential for prostate cells to proliferate in the absence of androgens. EPI-001 is active against castration-resistant prostate cancer (CRPC) in xenografts.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva De Mol et al.
ACS chemical biology, 11(9), 2499-2505 (2016-06-30)
Castration-resistant prostate cancer is the lethal condition suffered by prostate cancer patients that become refractory to androgen deprivation therapy. EPI-001 is a recently identified compound active against this condition that modulates the activity of the androgen receptor, a nuclear receptor
Raymond J Andersen et al.
Cancer cell, 17(6), 535-546 (2010-06-15)
Castration-recurrent prostate cancer (CRPC) is suspected to depend on androgen receptor (AR). The AF-1 region in the amino-terminal domain (NTD) of AR contains most, if not all, of the transcriptional activity. Here we identify EPI-001, a small molecule that blocked
Lucas J Brand et al.
Oncotarget, 6(6), 3811-3824 (2015-02-12)
The androgen receptor (AR) is a driver of prostate cancer (PCa) cell growth and disease progression. Therapies for advanced PCa exploit AR dependence by blocking the production or action of androgens, but these interventions inevitably fail via multiple mechanisms including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service