Amino Acid Sequence
Tyr-Glu-Val-His-His-Gln-Lys
General description
Amyloid beta aggregation leading to profibril formation and neurotoxicity is associated with Alzheimer′s disease. Several antibodies require the presence of amino acids 10-16 for Aβ detection. This peptide may be useful as a control in immunohistochemistry, ELISA and other antibody based analysis.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service