EP5895100007
Rotor for Eppendorf® 5920R Centrifuges
swing-bucket rotor, S-4x1000, aerosol-tight, to hold, 0.2 mL to 1,000 mL tubes and flasks (plate/tube buckets), with 4 × 1000 mL round buckets, with Eppendorf QuickLock® lid
About This Item
Recommended Products
feature
to hold 0.2 mL to 1,000 mL tubes and flasks (plate/tube buckets)
aerosol-tight
swing-bucket rotor
with 4 × 1000 mL round buckets
with Eppendorf QuickLock® lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5895100007 (Global)
parameter
3,428 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5920R
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)