Key Documents
BMSIPD9600E
Sigma-Aldrich
Benchmark BeadBlaster 96 Ball Mill Homogenizer
AC/DC input 220 V, Schuko plug
Synonym(s):
carbon dioxide incubator
About This Item
UNSPSC Code:
41101504
NACRES:
NB.73
feature
to hold Microplates, 15 & 50 mL tubes, microtubes and vials
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPD9600-E
parameter
1800 rpm speed
General description
The BeadBlaster 96 is an extremely versatile bead mill homogenizer that has applications in a variety of areas, including biological research, environmental testing, and industrial settings. Adapters are available for microplates, microtubes, 4ml vials and 15/50ml tubes. Grinding jars and media in a variety of materilas are also available. Wet homogenization, dry grinding and cryogrinding can all be performed with the BeadBlaster 96. The arced motion of the sample holders and powerful motor allow most samples to be processed in 1 minute or less. All parameters are digitally controlled through the large touch screen and 9 programs can be stored in memory. The safety cover lifts upward for easy access to the sample holders and automatically locks during operation. The wrap-around design of the clear acrylic lid allows for visualization during homogenization.
Features and Benefits
  • Accepts samples in microplates, tubes from 2ml to 50ml and grinding jars to 50ml.
  • Homogenize, wet or dry grind and cryogrind all in the same unit.
  • Large variety of adapters to accommodate a wide range of samples and sample containers.
Legal Information
BeadBlaster is a trademark of Benchmark Scientific, Inc.