Skip to Content
Merck
All Photos(1)

Key Documents

178497-M

Millipore

Apoptosis Activator VII, Apoptozole

The Apoptosis Activator VII, Apoptozole, also referenced under CAS 1054543-47-3, modulates Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Apoptosis Activator VII, Apoptozole, Hsp70 Inhibitor III, N-(4-Carboxamidobenzyl)-2-(3,5-bis-trifluoromethyl)-4,5-bis-(4-methoxyphenyl)-imidazole, N-(4-Carboxamidobenzyl)-2-(3,5-bis-trifluoromethyl)-4,5-bis-(4-methoxyphenyl)-imidazole, Hsp70 Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H24F6N3O3
CAS Number:
Molecular Weight:
624.55
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

ethanol: 5 mg/mL
DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(cc(c1)c2[n](c(c(n2)c5ccc(cc5)OC)c4ccc(cc4)OC)Cc3ccc(cc3)C(=O)N)C(F)(F)F

InChI

1S/C33H25F6N3O3/c1-44-26-11-7-20(8-12-26)28-29(21-9-13-27(45-2)14-10-21)42(18-19-3-5-22(6-4-19)30(40)43)31(41-28)23-15-24(32(34,35)36)17-25(16-23)33(37,38)39/h3-17H,18H2,1-2H3,(H2,40,43)

InChI key

ZIMMTPFXOMAJTQ-UHFFFAOYSA-N

General description

A cell-permeable imidazole compound that directly binds anti-apoptotic chaperone Hsc70 and its inducible homologue Hsp70 (Kd = 210 and 140 nM, respectively). Apoptozole is shown to effectively inhibit the proliferation of SK-OV-3, HCT-15, and A549 cancer cell lines (IC50 = 220, 250 and 130 nM, respectively) and induce 90% apoptotic cell death in P19 embryonic carcinoma and A549 lung cancer cultures after 12 h drug treatment (1 µM). Shown to suppress ubiquitination of mutant CFTR and promote membrane trafficking, and restore chloride channel activity.
A cell-permeable imidazole compound that directly binds anti-apoptotic chaperone Hsc70 and its inducible homologue Hsp70 (Kd = 210 and 140 nM, respectively). Apoptozole is shown to effectively inhibit the proliferation of SK-OV-3, HCT-15, and A549 cancer cell lines (IC50 = 220, 250 and 130 nM, respectively) and induce 90% apoptotic cell death in P19 embryonic carcinoma and A549 lung cancer cultures after 12 hours drug treatment (1 µM).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Cho, H.J., et al. 2012. J. Am. Chem. Soc.133, 20267.
Williams, D.R., et al. 2008. Angew. Chem. Int. Ed.47, 7466.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service