Skip to Content
Merck
All Photos(1)

Key Documents

1.67527

Supelco

Dip tube (PVDF) for nitric acid and sulfuric acid

pkg of 25 L (Combi-containers)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NB.51
Pricing and availability is not currently available.

packaging

pkg of 25 L (Combi-containers)

storage temp.

no temp limit

Related Categories

General description

in 25L combi container

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What are the thread dimensions for the dip tube and what are the accessories to use with the product for transferring?

    1 answer
    1. To perform a manual withdrawal using the PVDF dip tube product 1675270001, it would need the dispense head and either the hand or the foot pump, which are products 1.67501.0001, 9.67114.0000, and 1.67502.0001. The dip tube features an S56x4 outer thread and a G3/4 dust protection cap/stopper. The inner thread of the dip tube (NPT 1/4") is designed for the thread connection to the dispenser head 1.67501.0001 (PVDF) / 1.67500.0001 (PE).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service