Key Documents
Z178667
Hamilton® laboratory tubing clamp
fits vessels with rim thickness < 1/8 in., 12-18 gauge tubing size.
About This Item
UNSPSC Code:
41105108
NACRES:
NB.51
manufacturer/tradename
Hamilton
Related Categories
General description
Snap to rim of any laboratory vessel with rim thickness less than 1/8 in. to secure tubing from 12 to 18 gauge in size.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)