GF99172491
Alumina
tube, outside diameter 3.0 mm, length 1000 mm, wall thickness 0.7 mm
Synonym(s):
AL607060
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
tubes
manufacturer/tradename
Goodfellow 991-724-91
L
1000 mm
O.D.
3.0 mm
wall thickness
0.7 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service