Skip to Content
Merck
All Photos(1)

Documents

GF04189585

Stainless steel - AISI 316l

Non-Woven Fabric, Fe/Cr18%/Ni 10%/Mo 3%, weight.m-2 -50g, size 300 × 300 mm, thickness 0.060 mm

Synonym(s):

FF213720

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

form

fabric

manufacturer/tradename

Goodfellow 041-895-85

size

300 × 300 mm

thickness

0.060 mm

weight

50 gper meter squared

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service