BMSZ207AE
Benchmark Sprint 8 Plus Clinical Centrifuge with fixed angle rotor
to hold, 8 x 15 mL
Synonym(s):
Laboratory centrifuge, lab centrifuge, clinical centrifuge, blood centrifuge, urine centrifuge, 15ml centrifuge, Low speed centrifuge
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
AC/DC input
230 V, EuroPlug
feature
refrigerated: no
to hold 8 x 15 mL
manufacturer/tradename
Benchmark Scientific Z207-A-E
parameter
6800 rpm speed
Looking for similar products? Visit Product Comparison Guide
General description
Unlike traditional clinical centrifuges the increased speed capability (4445xg) of the Sprint TM 8 PLUS allows for the separation of serum or plasma in as little as 23 minutes. Additional features include:
A large backlit ,liquid crystal display
High capacity rotor, 8 tubes up to 15ml (conical or round bottom)
Unique EZScroll touch pad
Timer up to 99hr99mins. / continuous
Storage of commonly used runtimes
Control of speed in both rpm and rcf
Adjustable acceleration and deceleration levels
Safety imbalance detection system
Momentary operation Electrical lid lock
Ideal for rapid production of serum and plasma
A large backlit ,liquid crystal display
High capacity rotor, 8 tubes up to 15ml (conical or round bottom)
Unique EZScroll touch pad
Timer up to 99hr99mins. / continuous
Storage of commonly used runtimes
Control of speed in both rpm and rcf
Adjustable acceleration and deceleration levels
Safety imbalance detection system
Momentary operation Electrical lid lock
Ideal for rapid production of serum and plasma
Application
Centrifugation of clinical samples including urine and blood. Ideal for production of PPP and PRP.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service