description
Electrode coating: Au 3 µin
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
concentration
substrate (FR4)
electrode surface area
1 cm2
size
49 × 16 × 0.5 mm
weight
0.767 g
greener alternative category
General description
Our flat square electrode has a surface area of 1cm2, it is coated with gold, chemically stable, biocompatible, and durable. Its main applications include: Energy material characterization; Sensors; Energy storage devices; Actuators; Material synthesis.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)