Skip to Content
Merck
All Photos(1)

Key Documents

685003

Sigma-Aldrich

5-Fluoro-2-methylbenzaldehyde

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7FO
CAS Number:
Molecular Weight:
138.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

96%

form

liquid

refractive index

n20/D 1.5244

density

1.1446 g/mL at 25 °C

SMILES string

Cc1ccc(F)cc1C=O

InChI

1S/C8H7FO/c1-6-2-3-8(9)4-7(6)5-10/h2-5H,1H3

InChI key

MBOXPKNOGZJXPK-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.1 °F - closed cup

Flash Point(C)

76.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service