MilliporeSigma
  • The effect of cocoa and polydextrose on bacterial fermentation in gastrointestinal tract simulations.
The effect of cocoa and polydextrose on bacterial fermentation in gastrointestinal tract simulations.
Bioscience, biotechnology, and biochemistry (2007-08-11)
Harri Mäkivuokko, Hannele Kettunen, Markku Saarinen, Tatsuya Kamiwaki, Yasuhiro Yokoyama, Julian Stowell, Nina Rautonen
ABSTRACT
Effects of cocoa mass and supplemented dietary fiber (polydextrose) on microbial fermentation were studied by combining digestion simulations of stomach and small intestine with multi-staged colon simulations. During the four phases of digestion, concentrations of available soluble proteins and reducing sugars reflected in vivo absorption of nutrients in small intestine. In colon simulation vessels, addition of polydextrose to digested cocoa mass significantly increased concentrations of total short-chain fatty acids and butyric acid, from 103 to 468 mM (P<0.01) and from 12 to 22 mM (P<0.01), respectively. Long-chain fatty acid concentrations (decreasing from 1,222 to 240 mM) were mainly affected by the presence of digested cocoa mass. Cocoa mass with or without polydextrose addition significantly decreased production of cadaverine (P<0.02) and branched-chain fatty acids compared to control during colon simulations. Results indicate beneficial effects on metabolism of colonic microbiota after digestion of cocoa mass, and even more so with polydextrose addition.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid