Merck
Global profiling of dynamic protein palmitoylation.
Nature methods (2011-11-08)
Brent R Martin, Chu Wang, Alexander Adibekian, Sarah E Tully, Benjamin F Cravatt
ABSTRACT
The reversible thioester linkage of palmitic acid on cysteines, known as protein S-palmitoylation, facilitates the membrane association and proper subcellular localization of proteins. Here we report the metabolic incorporation of the palmitic acid analog 17-octadecynoic acid (17-ODYA) in combination with stable-isotope labeling with amino acids in cell culture (SILAC) and pulse-chase methods to generate a global quantitative map of dynamic protein palmitoylation events in cells. We distinguished stably palmitoylated proteins from those that turn over rapidly. Treatment with a serine lipase-selective inhibitor identified a pool of dynamically palmitoylated proteins regulated by palmitoyl-protein thioesterases. This subset was enriched in oncoproteins and other proteins linked to aberrant cell growth, migration and cancer. Our method provides a straightforward way to characterize global palmitoylation dynamics in cells and confirms enzyme-mediated depalmitoylation as a critical regulatory mechanism for a specific subset of rapidly cycling palmitoylated proteins.
Sigma-Aldrich
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
17-Octadecynoic acid, ≥95% (GC)