Merck
Total synthesis of (-)-reveromycin a.
Organic letters (2004-08-28)
Mariana El Sous, Danny Ganame, Peter A Tregloan, Mark A Rizzacasa
ABSTRACT
The asymmetric total synthesis of (-)-reveromycin A is described. The key steps involved a Lewis acid catalyzed inverse electron demand hetero-Diels-Alder reaction followed by hydroboration/oxidation to afford the spiroketal core 4 in a highly stereoselective manner and introduction of the C18 hemisuccinate by high-pressure acylation.
Sigma-Aldrich
Reveromycin A, solid