Merck
  • Initiation, growth and cryopreservation of plant cell suspension cultures.
Initiation, growth and cryopreservation of plant cell suspension cultures.
Nature protocols (2011-06-04)
Natali R Mustafa, Ward de Winter, Frank van Iren, Robert Verpoorte
ABSTRACT
Methods described in this paper are confined to in vitro dedifferentiated plant cell suspension cultures, which are convenient for the large-scale production of fine chemicals in bioreactors and for the study of cellular and molecular processes, as they offer the advantages of a simplified model system for the study of plants when compared with plants themselves or differentiated plant tissue cultures. The commonly used methods of initiation of a callus from a plant and subsequent steps from callus to cell suspension culture are presented in the protocol. This is followed by three different techniques for subculturing (by weighing cells, pipetting and pouring cell suspension) and four methods for growth measurement (fresh- and dry-weight cells, dissimilation curve and cell volume after sedimentation). The advantages and disadvantages of the methods are discussed. Finally, we provide a two-step (controlled rate) freezing technique also known as the slow (equilibrium) freezing method for long-term storage, which has been applied successfully to a wide range of plant cell suspension cultures.
Sigma-Aldrich
EZMix N-Z-Amine® A, Casein enzymatic hydrolysate, from bovine milk