
Hematoxylin cryst. (C.I. 75290)

Synonym(s):
Hematoxylin cryst. (C.I. 75290)
Empirical Formula (Hill Notation):
C16H14O6
CAS Number:
Molecular Weight:
302.28
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing