Key Documents
509485
SGE Gas Tight Syringes, Luer Lock
500F-LL-GT, volume 500 μL, needle size (not included)
Synonym(s):
SYRINGE SGE 500UL, LUER LOCK
About This Item
UNSPSC Code:
41122001
material
glass syringe
needle size
(not included)
feature
Fixed Luer Lock
packaging
pkg of 1 ea
manufacturer/tradename
SGE 007230
volume
500 μL
General description
SGE gas tight syringe without a needle, Model: 500F-LL-GT (volume 0.5 mL)
  • Made of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
  • Autoclavable syringes but the plunger should be removed.
  • Features an interference fit between the plunger tip and the barrel which enhances the gas sealing properties of the syringe.
  • Maximum inertness for liquid or gas samples.
  • Suitable for manual dispenser/diluter applications.
  • High accuracy of dispensed volume.
Choose from one of the most recent versions:
Certificates of Analysis (COA)