27134
Vials, screw top with solid green Thermoset cap with PTFE liner, preassembled, pkg of 100
volume 2 mL, clear glass vial (standard opening - 4.6 mm), O.D. × H × I.D. 12 mm × 32 mm × 4.6 mm, thread for 8-425
Synonym(s):
PK100 2ML CLEAR VIAL,SCREW TOP, SOLID CAP W/PTFE LINER
About This Item
Recommended Products
material
F217/PTFE cap liner
PTFE liner
clear glass vial (standard opening - 4.6 mm)
green melamine resin cap (solid-top)
feature
closure type green melamine solid-top cap
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H × I.D.
12 mm × 32 mm × 4.6 mm
volume
2 mL
fitting
thread for 8-425
Looking for similar products? Visit Product Comparison Guide
General description
related product
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service