Skip to Content
Merck
All Photos(11)

Key Documents

ZRB1677

Sigma-Aldrich

Anti-Wilms Tumor Protein Antibody, clone 2O18 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

WT33

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2O18, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 49.19 kDa
observed mol wt ~54 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey, mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... WT1(7490)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2O18 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Wilms Tumor Protein (WT33). It targets an epitope within 23 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the N-terminal half of human Wilms Tumor Protein.

Application

Quality Control Testing

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Wilms Tumor Protein in HEK293 cell lysate.

Tested applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected Wilms Tumor Protein in OVCAR-3 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Wilms Tumor Protein (WT33) in OVCAR-3 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Wilms Tumor Protein (WT33) in human kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Wilms tumor protein (UniProt: P19544; also known as WT33) is encoded by the WT1 gene (Gene ID: 7490) in human. It serves as a transcription factor that plays an important role in cellular development and cell survival. It is expressed in the kidney and a subset of hematopoietic cells. It regulates the expression of numerous target genes, including EPO. Wilms tumor protein plays an essential role for development of the urogenital system and has a tumor suppressor as well as an oncogenic role in tumor formation. Eight different isoforms of Wilms tumor protein have been described that are produced by alternative splicing. Isoforms lacking the KTS motif act as transcription factors while isoforms containing the KTS motif may bind mRNA and play a role in mRNA metabolism or splicing. Isoform 1 has lower affinity for DNA and can bind RNA. The N-terminal region of Wilms tumor protein contains a proline-rich region (aa 27-83) involved in transcriptional regulation, self-association, and RNA recognition, while its C-terminal region contains four C2H2-type zinc fingers (aa 323-347, 353-377, 383-405, 414-438) that mediate DNA and RNA binding. Its zinc finger domain can bind to GC-rich sequences, such as the EGR-1 consensus sequence (5 -GCG(T/G)GGGCG-3 ), the WTE motif (5′-GCGTGGGAGT-3′), or (TCC)n motif. Mutations in WT1 gene are reported to cause Wilms tumors characterized by embryonal malignancy of the kidney. Some mutations are reported to cause nephrotic syndrome 4, which causes severe proteinuria, resulting in complications such as hypoalbuminemia, hyperlipidemia and edema. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service