Z721573
AeraSeal™
sterile
Synonym(s):
breathable plate seal, permeable rayon film, piercable plate seal, tissue culture plate seal
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
Rayon film
sterility
sterile
feature
piercable
packaging
pack of 50 ea
manufacturer/tradename
Excel Scientific BS-25
technique(s)
tissue culture: suitable
W × L
8.26 cm × 14.29 cm
General description
A 140 mm thick hydrophobic porous sealing film with medical-grade adhesive for tissue culture plates, bio-blocks, and 96-well plates where gas exchange is necessary for cell or bacterial growth. AeraSealÔ sealing films minimize cross-contamination, spillage and evaporation. They allow uniform air and CO2 exchange for all wells, unlike plate lids which favor exchange for wells near the plate edges. Dimensions: 82.6 by 142.9 mm for standard-size tissue culture plate
Note: AeraSealÔ films are suitable for hypoxic culture conditions
- non-cytotoxic, highly gas permeable
- easily pierceable with pipet tips or pipets for sample recovery
- recommended for temperatures from -20 °C to +80 °C
Note: AeraSealÔ films are suitable for hypoxic culture conditions
Application
- AeraSeal films have been used for the covering of 96-well opaque white plates to incubate bacterial cells.
- It has been used for the sealing of 96 deep-well plate for culturing of yeast cells.
- It has been used for the sealing of 96-well microtitre plate to separate mites.
Legal Information
AeraSeal is a trademark of Excel Scientific, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service