Key Documents
Z657883
Hettich® centrifuge adapters
for ROTINA 38/38R and UNIVERSAL 320/320R, Adapter for 50 mL tube 34x100 mm
About This Item
UNSPSC Code:
41103911
NACRES:
NB.22
feature
Holds 50 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Hettich 5233
technique(s)
centrifugation: suitable
compatibility
for use with 1427 (bucket)
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)