TMO374026
Nunc® Coded Cryobank Vials in black low rack
polypropylene, capacity 0.5 mL, self-standing natural, sterile, red cap, case of 960 ea
Synonym(s):
coded cryobank vial storage, cryostorage
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121806
NACRES:
NB.11
Recommended Products
material
polypropylene
red cap
self-standing natural
sterility
sterile
packaging
case of 960 ea
manufacturer/tradename
Nunc 374026
technique(s)
cryopreservation: suitable
capacity
0.5 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Related applications: Cell Culture Plastics
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service