Skip to Content
Merck
All Photos(1)

Key Documents

R7026

Sigma-Aldrich

PCR 96 Well rack

natural

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122804
NACRES:
NB.22
Pricing and availability is not currently available.

material

polypropylene

feature

Holds 96 x 0.2 mL tubes

manufacturer/tradename

Heathrow Scientific HS23461B

technique(s)

PCR: suitable

size

127x99x30 mm

color

natural

Looking for similar products? Visit Product Comparison Guide

General description

Sturdy rack for working with 0.2 mL tubes. Accommodates the standard 8 or 12 tube strips. Perfect for pre- or post-PCR sampling applications. The tube wells are easily identified with imprinted numbers and letters. The rack features a removable hinged lid.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service