Skip to Content
Merck
All Photos(1)

Key Documents

L1417

Sigma-Aldrich

15(S)-Latanoprost solution

≥95%

Synonym(s):

(5Z,9α,11α,15S)-9,11,15-Trihydroxy-17-phenyl-18,19,20-trinor-prost-5-en-1-oate isopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H40O5
Molecular Weight:
432.59
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95%

form

liquid

concentration

10 mg/mL in methyl acetate

storage temp.

−20°C

SMILES string

CC(C)OC(=O)CCC\C=C/C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1CC[C@H](O)CCc2ccccc2

InChI

1S/C26H40O5/c1-19(2)31-26(30)13-9-4-3-8-12-22-23(25(29)18-24(22)28)17-16-21(27)15-14-20-10-6-5-7-11-20/h3,5-8,10-11,19,21-25,27-29H,4,9,12-18H2,1-2H3/b8-3-/t21-,22-,23-,24+,25-/m1/s1

InChI key

GGXICVAJURFBLW-SCTZCWPJSA-N

Linkage

15(S)-Latanoprost is the less active C-15 isomer of latanoprost where the C-15 hydroxyl group is inverted relative to its natural (α) position in a prostaglandin such as PGF. The 15(S)-isomer of latanoprost occurs as an impurity of between 0.1 and 1% in most commercial preparations of latanoprost.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

3.2 °F - closed cup

Flash Point(C)

-16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service