HS1421B
Weigh Boat
Medium, square blue polystyrene, pk of 500 ea
Synonym(s):
weigh boat, weigh dish, weighing dish, weighing pan
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.35
Recommended Products
material
square blue polystyrene
size
medium
packaging
pk of 500 ea
manufacturer/tradename
Heathrow Scientific HS1421B
L × W × H
(85 mm) × 84 mm × 24 mm
color
blue
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Features and Benefits
- biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
- temperature range -10 to 70C
- disposable
- shallow with a wide, flat bottom to resist tipping
- rounded corners simplify transfer
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service