Skip to Content
Merck
All Photos(2)

Key Documents

GBL204960

Grace Bio-Labs ProPlate MP microtiter plate superstructure

size 96 wells, black polystyrene, adhesive

Synonym(s):

bottomless adhesive microtiter plate, square well microtitier plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600
NACRES:
NB.22

material

black polystyrene
square wells

sterility

non-sterile

packaging

pack of 1 ea

manufacturer/tradename

Grace Bio-Labs 204960

W × L

85.3 mm × 127.6 mm

size

96 wells

well volume

350 μL

color

none

Looking for similar products? Visit Product Comparison Guide

General description

N/A

Legal Information

ProPlate MP is a trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service