Skip to Content
Merck
All Photos(1)

Key Documents

AXYEV100R

Pipette tips for Tecan® EVO/ Caliper Zephyr® Automated Liquid Handling System

racked, capacity 100 μL, pkg of 5x960tips/cs, clear

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02

material

clear

sterility

non-sterile

feature

DNA free
RNase free
barrier: no
endotoxin free
filter: no

manufacturer/tradename

Axygen Inc EV-100-R
Corning

packaging

pkg of 5x960tips/cs

capacity

100 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Tecan is a registered trademark of Tecan Group AG
Zephyr is a registered trademark of Caliper Life Sciences, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service