PHR3367
Ticlopidine hydrochloride
certified reference material, pharmaceutical secondary standard
Synonym(s):
5-(o-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to Ph. Eur. T1423000
traceable to USP 1667144
packaging
pkg of 300 mg
application(s)
pharmaceutical small molecule
storage temp.
2-30°C
SMILES string
Cl.Clc1ccccc1CN2CCc3sccc3C2
InChI
1S/C14H14ClNS.ClH/c15-13-4-2-1-3-11(13)9-16-7-5-14-12(10-16)6-8-17-14;/h1-4,6,8H,5,7,9-10H2;1H
InChI key
MTKNGOHFNXIVOS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service