Skip to Content
Merck
All Photos(1)

Key Documents

43753

Millipore

Legionella BCYE Supplement without L-cysteine

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.85
Pricing and availability is not currently available.

Agency

according to ISO 11731:2017

Quality Level

sterility

sterile

form

liquid

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Legionella spp.

Application

A chemical enrichment supplement without L-Cysteine for presumptive identification of Legionella species.

Components

(per vial, sufficient for 450 ml medium)
ACES Buffer/Potassium hydroxide 5g
Ferric pyrophosphate, soluble 0.125g
α-Ketoglutarate 0.5g
Distilled water 50ml

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service