MABN2434M
Anti-Retinoschisin Antibody, clone 3R10
clone 3R10, from mouse
Synonym(s):
X-linked juvenile retinoschisis protein, RS1-3R10
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
3R10, monoclonal
species reactivity
human, mouse
packaging
antibody small pack of 25 μg
technique(s)
immunofluorescence: suitable
western blot: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... RS1(6247)
General description
Specificity
Immunogen
Application
Western Blotting Analysis: A representative lot detected Retinoschisin in Western Blotting applications (Min, S.H., et. al. (2005). Mol Ther. 12(4):644-51; Wu, W.W., et. al. (2003). J Biol Chem. 278(30):28139-46).
Neuroscience
Quality
Western Blotting Analysis: 2 µg/mL of this antibody detected Retinoschisin in 10 µg of mouse retinal tissue lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service