Skip to Content
Merck
All Photos(1)

Key Documents

8.41398

Sigma-Aldrich

4-Methoxystyrene

for synthesis

Synonym(s):

4-Methoxystyrene, 4-Vinylanisole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O
CAS Number:
Molecular Weight:
134.18
MDL number:
UNSPSC Code:
12352112
EC Index Number:
211-298-9
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥97.5% (GC)

form

liquid

transition temp

flash point 76 °C

density

1.002 g/cm3 at 20 °C

SMILES string

O(C)c1ccc(cc1)C=C

InChI

1S/C9H10O/c1-3-8-4-6-9(10-2)7-5-8/h3-7H,1H2,2H3

InChI key

UAJRSHJHFRVGMG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.995 - 1.001
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service