Skip to Content
Merck
All Photos(1)

Key Documents

8.18677

Sigma-Aldrich

Lauraldehyde

for synthesis

Synonym(s):

Lauraldehyde, Dodecanal, Dodecylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CHO
CAS Number:
Molecular Weight:
184.32
MDL number:
UNSPSC Code:
12352114
EC Index Number:
203-983-6
NACRES:
NA.22

vapor pressure

<300 hPa ( 50 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

200 °C

potency

23000 mg/kg LD50, oral (Rat)

bp

237-239 °C/1013 hPa

mp

11-13 °C

transition temp

flash point 101 °C

density

0.83 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

O=CCCCCCCCCCCC

InChI

1S/C12H24O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h12H,2-11H2,1H3

InChI key

HFJRKMMYBMWEAD-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.830 - 0.840
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service