Skip to Content
Merck
All Photos(1)

Key Documents

8.18530

Sigma-Aldrich

4-Cyanobenzaldehyde

for synthesis

Synonym(s):

4-Cyanobenzaldehyde, 4-Formylbenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(OHC)C6H4CN
CAS Number:
Molecular Weight:
131.13
MDL number:
UNSPSC Code:
12352117
EC Index Number:
203-267-3
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

bp

133 °C/16 hPa

mp

100-103 °C

storage temp.

2-30°C

SMILES string

N#Cc1ccc(cc1)C=O

InChI

1S/C8H5NO/c9-5-7-1-3-8(6-10)4-2-7/h1-4,6H

InChI key

WZWIQYMTQZCSKI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 97 °C
Melting range (upper value): ≤ 101 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service