Skip to Content
Merck
All Photos(1)

Documents

07-1511

Sigma-Aldrich

Anti-phospho-WAVE2 (Ser308) Antibody

Synonym(s):

Wiskott-Aldrich syndrome protein family member 2, WASP family protein member 2, Protein WAVE-2, Verprolin homology domain-containing protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody product type

primary antibodies

Quality Level

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark A Munger et al.
Nanomedicine : nanotechnology, biology, and medicine, 10(1), 1-9 (2013-07-03)
Human biodistribution, bioprocessing and possible toxicity of nanoscale silver receive increasing health assessment. We prospectively studied commercial 10- and 32-ppm nanoscale silver particle solutions in a single-blind, controlled, cross-over, intent-to-treat, design. Healthy subjects (n=60) underwent metabolic, blood counts, urinalysis, sputum
Yanan Wang et al.
Nature communications, 14(1), 3541-3541 (2023-06-16)
The RAC1-WAVE-Arp2/3 signaling pathway generates branched actin networks that power lamellipodium protrusion of migrating cells. Feedback is thought to control protrusion lifetime and migration persistence, but its molecular circuitry remains elusive. Here, we identify PPP2R1A by proteomics as a protein
ERK-MAPK drives lamellipodia protrusion by activating the WAVE2 regulatory complex.
Mendoza, MC; Er, EE; Zhang, W; Ballif, BA; Elliott, HL; Danuser, G; Blenis, J
Molecular Cell null
Sascha Gromnitza et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 32(3), 1665-1676 (2017-11-23)
Podocyte malfunction is central to glomerular diseases and is marked by defective podocyte intercellular junctions and actin cytoskeletal dynamics. Podocytes share many morphologic features with neurons, so that similar sets of proteins appear to regulate cell process formation. One such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service