Skip to Content
Merck
All Photos(1)

Key Documents

GF75473062

Chromium-nickel alloy, Cr50Ni50

powder, 420micron max. particle size, 200g

Synonym(s):

CR026010

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141709
NACRES:
NA.23

form

powder

manufacturer/tradename

Goodfellow 754-730-62

weight

200 g

max. part. size

420 μm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service