GF06217006 (0.05 micron) is in the beta phase. The following products are all in the alpha phase: GF29667190 (0.1-1 micron), GF10020339, GF09680160, GF08399391, GF49923771, GF48263014, GF40471455, and GF32877445.
GF06217006
Silicon carbide
powder, max. particle size 0.05 micron, weight 100 g, purity 99%
Synonym(s):
SI516022
About This Item
Recommended Products
Assay
99%
form
powder
manufacturer/tradename
Goodfellow 062-170-06
weight
100 g
max. part. size
0.05 μm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Are the Goodfellow silicon carbide products that are offer in the alpha phase or beta phase? The specific products are: GF06217006 (0.05 micron), GF29667190 (0.1-1 micron), GF10020339, GF09680160, GF08399391, GF49923771, GF48263014, GF40471455, and GF32877445.
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service