DWKBPM2000P
Azlon Tapered Beakers with Moulded Graduations
polypropylene, capacity 2000 mL
Synonym(s):
BPM2000P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
Recommended Products
material
polypropylene
packaging
pkg of 1 ea
manufacturer/tradename
Azlon
storage condition
room temperature
assay range
accuracy: 200 mL
capacity
2000 mL
Looking for similar products? Visit Product Comparison Guide
Application
Excellent solution for measuring and dispensing laboratory solutions and reagents
Features and Benefits
Autoclavable at 121°C and complies with ISO 7056
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service