Key Documents
BR65815
BRAND® desiccator
size 250 mm
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
Pricing and availability is not currently available.
description
lid PC, base & desiccant tray PP
lid PC, base & desiccant tray PP
packaging
pack of 1 ea
manufacturer/tradename
BRAND 65815
size
250 mm
Related Categories
General description
Polycarbonate lid. PP base and desiccant tray. Venting stopper with non-return valve in lid. Seal between lid and base: polychlorobutadiene rubber (CK) O-ring, compressed by vacuum. Light-weight and easy to handle.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)