Key Documents
BAF378633000
SP Bel-Art® Colony Counter System with Mini LED Light Box and Magnifier
Includes Hand-Held Colony Counter (F37862-0002), Mini LED Light Box (F37864-3000) and Mini Magnifier (F37865-0000)
About This Item
UNSPSC Code:
41104510
NACRES:
NC.56
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37863-3000
Related Categories
General description
A complete, effective and accurate way to count colonies of bacteria or other microorganisms grown on an agar plate. Provides equipment for proper lighting, magnification and tracking of colonies Includes Hand-Held Colony Counter (F37862-0002), Mini LED Light Box (F37864-3000) and Mini Magnifier (F37865-0000)
Features and Benefits
  • Hand-Held Colony Counter marks, counts, and gives an audible and visual alert each time a count is registered
  • Mini LED Light Box enhances viewing of transparent and semi-transparent objects
  • Mini Magnifier offers 1.75X magnification
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)