Skip to Content
Merck
All Photos(1)

Key Documents

PHL83536

Rhein 8-glucoside

phyproof® Reference Substance

Synonym(s):

Rhein 8-O-β-D-glucopyranoside, 8-Glucosylrhein, Glucorhein, Rhein 8-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18O11
CAS Number:
Molecular Weight:
446.36
Beilstein:
1359724
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

2-8°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)Oc2c3c(ccc2)C(=O)c4c(c(cc(c4)C(=O)O)O)C3=O

InChI

1S/C21H18O11/c22-6-12-16(25)18(27)19(28)21(32-12)31-11-3-1-2-8-14(11)17(26)13-9(15(8)24)4-7(20(29)30)5-10(13)23/h1-5,12,16,18-19,21-23,25,27-28H,6H2,(H,29,30)/t12-,16-,18+,19-,21-/m1/s1

InChI key

WYKUTTFFZMQCGO-HTRBZNBPSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaochao Zhang et al.
Current microbiology, 78(1), 323-328 (2020-11-01)
Dental caries is the most frequent biofilm-related human infectious disease in the oral cavity. Streptococcus mutans is one of the primary etiological agents of dental caries. The aim of our study was to investigate the effects of rhein-8-O-β-D-glucopyranoside (Rg) on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service