Z744280
Wilmad® Universal Jet Washer for EPR and NMR Sample Tubes
tube O.D. 2.5-5 mm, inner joint: S/T 24/40
Synonym(s):
EPR tube washer, NWR tube washer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121705
NACRES:
NB.15
Recommended Products
Joint
inner joint: S/T24/40
packaging
pack of 1 ea
manufacturer/tradename
Wilmad WG-7200-J1
tube O.D.
2.5-5 mm
Looking for similar products? Visit Product Comparison Guide
General description
Universal Jet NMR Tube Washer can be used for any length NMR sample tube by simply adjusting the included flexible PTFE tubing. The washer head is then affixed to a 24/40 joint filter flask (with vacuum sidearm) and the tubing is inserted into a washing solvent reservoir. An inverted sample tube is placed over the PTFE tubing, a vacuum is applied to the flask, sample tube is pressed against the rubber gasket to form an air-tight seal that starts the solvent flow. By lifting the PTFE tubing out of the solvent reservoir, the sample tube can be air-dried.
Application
NMR
Features and Benefits
For any length NMR sample tube
Legal Information
Wilmad is a registered trademark of SP Industries
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service